missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IL-1 beta/IL-1F2 DuoSet ELISA

Product Code. p-4754778
Click to view available options
Packaging Quantity:
15 Plates
5 Plates
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 30021398

Brand: R&D Systems™ DY401

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse IL-1 beta/IL-1F2 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse IL-1 beta/IL-1F2 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse IL-1 beta/IL-1F2.
TRUSTED_SUSTAINABILITY

Specifications

Product Type ELISA Development Kit
Quantity 1 Kit
Species Mouse
Conjugate Biotin
Assay Range 15.60 to 1000 pg/mL
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
For Use With (Application) ELISA
Gene ID (Entrez) 16176
Synonym catabolin, IL-1, IL1 beta, IL-1 beta, IL1B, IL-1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta
Packaging Quantity 15 Plates
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.