missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Aprotinin
Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
Description
Cell dissociation reagent isolated from bovine lung. Lyophilized.
Specifications
Specifications
| Content And Storage | 4°C |
| Form | Powder |
| Product Type | Trypsin inhibitor |
| Sterility | Sterile |
| pH | 5 to 7 |
| Quantity | 10 mg |
| Source | Bovine lung |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction