missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Hyaluronidase, Bovine Testes

Product Code. 11343929 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
This item is not returnable. View return policy

Product Code. 11343929

Brand: MP Biomedicals™ 0210074090

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Bovine Testes

Hyaluronidase, [37326-33-3]

  • Hyaluronate Glycanohydrolase
  • Activity: 300 USP units/mg min.
  • Lyophilized powder

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≥300 U/mg
Color Tan
pH 4.5 to 6
For Use With (Application) Drug dispersion and delivery
Content And Storage -20°C
Source Bovine testes
Quantity 500 mg
Product Type Hyaluronidase
Molecular Weight (g/mol) ≈55
Inhibitors Fe2+ and Fe3+ are inhibitory as are Mn2+ and Cu2+
Form Lyophilized Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.