Learn More
MP Biomedicals™ MT Buffer
MT Buffer is a lysis solution, carefully developed to protect and solubilize nucleic acids and proteins upon cell lysis.
Brand:  MP Biomedicals™ 116511202
Product Code. 12877860
  • 48.05 € / 8mL
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • MT Buffer is a lysis solution component included in the FastRNAPro Soil Direct and Indirect Kits, the FastDNA Spin Kit for Feces, and the FastDNA 50 mL Spin Kit for Soil.
  • Used as a stabilization agent during sample lysis.
  • Homogenization takes place in the presence of MT Buffer, this reagent is carefully developed to protect and solubilize nucleic acids and proteins upon cell lysis.
  • Thus allowing extraction of genomic DNA with minimal RNA contamination.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Feces, Soil, Wastewater, Environmental Water, Sludge
Sample preparation, sample lysis, nucleic acid isolation and purification, molecular biology, gene expression, microbiological analysis, gene function, gene regulation
8 mL
Microorganisms
FastPrep-24, FastPrep 120, FastPrep-96, SuperFastPrep-1
MT Buffer
Up to 500 mg Per Prep
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MP Biomedicals™ MT Buffer > MT Buffer; Quantity: 8 mL
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.