Learn More
MRPL17 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-18407JF646
Product Code. 18577688
  • 663.00 € / 0.10mL
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
This item is not returnable. View return policy
Description
Description
MRPL17 Polyclonal antibody specifically detects MRPL17 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MRPL17
Polyclonal
Western Blot
L17mt, LIP2, LYST-interacting protein 2, LYST-interacting protein LIP2, mitochondrial ribosomal protein L17, MRP-L17, MRP-L26, RPL17L, RPML26,39S ribosomal protein L17, mitochondrial
Recognizes region between residues Between Between 25 and 75..
0.1 mL
Stem Cell Markers
63875
Store at 4°C in the dark.
IgG
Western Blot
Janelia Fluor 646
50mM Sodium Borate
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MRPL17 Antibody, Janelia Fluor™ 646, Novus Biologicals™ > 0.1 mL; Janelia Fluor 646