Learn More
IKA™ MX.PVDF Sample Mixers
MX.PVDF Set of 3 sample mixers made of polyvinylidene difluoride, each with a different mixing type.
Brand:  IKA™ 0020103093
Product Code. 17583636
  • 176.00 € / Each
Estimated Shipment: 14-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Set of 3 sample mixers made of polyvinylidene difluoride, each with a different mixing type.
  • The package includes:
    • T shape mixing chip * 1
    • Y shape mixing chip * 1
    • Interrupted flow mixing chip * 1.
    • All three mixing chips are made of polyvinylidene difluoride (PVDF).
  • Scope of delivery
    • T shape mixing chip, PVDF
Specifications
Specifications
Polyvinylidene Difluoride
T Shape Mixing Chip, Y Shape Mixing Chip, Interrupted Flow Mixing Chip