missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ MYL3 (Human) Recombinant Protein
Human MYL3 full-length ORF ( AAH09790, 1 a.a. - 195 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00004634-P01.10ug
This item is not returnable.
View return policy
Description
- Sequence: MAPKKPEPKKDDAKAAPKAAPAPAPPPEPERPKEVEFDASKIKIEFTPEQIEEFKEAFMLFDRTPKCEMKITYGQCGDVLRALGQNPTQAEVLRVLGKPRQEELNTKMMDFETFLPMLQHISKNKDTGTYEDFVEGLRVFDKEGNGTVMGAELRHVLATLGERLTEDEVEKLMAGQEDSNGCINYEAFVKHIMSS
Specifications
AAH09790 | |
myosin, light chain 3, alkali; ventricular, skeletal, slow | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MYL3 | |
GST |
4634 | |
Wheat germ expression system | |
10 μg | |
CMH8, MLC1SB, MLC1V, VLC1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |