missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-D-leucine, 99%, Thermo Scientific Chemicals

Product Code. 11446080
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19764-30-8
Molecular Formula C8H15NO3
Molecular Weight (g/mol) 173.21
MDL Number MFCD00066069
InChI Key WXNXCEHXYPACJF-JLDDOWRYNA-N
Synonym n-acetyl-d-leucine, ac-d-leu-oh, d-leucine, n-acetyl, n-acethy-d-leucine, n-acetyl-r-leucine, unii-91wu82ga22, 2,n-acetyl-d-leucine, r-2-acetamido-4-methylpentanoic acid, 2r-2-acetamido-4-methylpentanoic acid, acetylleucine, d
PubChem CID 1241420
IUPAC Name (2R)-2-acetamido-4-methylpentanoic acid
SMILES CC(C)C[C@@H](NC(C)=O)C(O)=O

Specifications

Formula Weight 173.21
Percent Purity 99%
Quantity 1 g
Chemical Name or Material N-Acetyl-D-leucine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'