missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals N-Acetyl-L-alanine, 96%
Chemical Identifiers
CAS | 97-69-8 |
---|---|
Molecular Formula | C5H9NO3 |
Molecular Weight (g/mol) | 131.131 |
MDL Number | MFCD00063132 |
InChI Key | KTHDTJVBEPMMGL-VKHMYHEASA-N |
Synonym | n-acetyl-l-alanine, ac-ala-oh, acetylalanine, acetyl-l-alanine, n-acetylalanine, 2-acetamidopropionic acid, l-n-acetylalanine, n-acetyl-s-alanine, l-alanine, n-acetyl, 2s-2-acetamidopropanoic acid |
PubChem CID | 88064 |
ChEBI | CHEBI:40992 |
IUPAC Name | (2S)-2-acetamidopropanoic acid |
SMILES | CC(C(=O)O)NC(=O)C |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
97-69-8 | |
131.131 | |
KTHDTJVBEPMMGL-VKHMYHEASA-N | |
88064 | |
(2S)-2-acetamidopropanoic acid |
C5H9NO3 | |
MFCD00063132 | |
n-acetyl-l-alanine, ac-ala-oh, acetylalanine, acetyl-l-alanine, n-acetylalanine, 2-acetamidopropionic acid, l-n-acetylalanine, n-acetyl-s-alanine, l-alanine, n-acetyl, 2s-2-acetamidopropanoic acid | |
CHEBI:40992 | |
CC(C(=O)O)NC(=O)C |
Specifications
122°C to 128°C | |
C5H9NO3 | |
1722932 | |
KTHDTJVBEPMMGL-VKHMYHEASA-N | |
(2S)-2-acetamidopropanoic acid | |
88064 | |
131.13 | |
−64° (c=2 in Water) |
97-69-8 | |
MFCD00063132 | |
n-acetyl-l-alanine, ac-ala-oh, acetylalanine, acetyl-l-alanine, n-acetylalanine, 2-acetamidopropionic acid, l-n-acetylalanine, n-acetyl-s-alanine, l-alanine, n-acetyl, 2s-2-acetamidopropanoic acid | |
CC(C(=O)O)NC(=O)C | |
131.131 | |
CHEBI:40992 | |
96% | |
N-Acetyl-L-alanine |
Safety and Handling
missing translation for 'einecsNumber' : 202-602-0
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only