missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-L-histidine monohydrate

48.90 € - 321.00 €

Chemical Identifiers

CAS 39145-52-3
Molecular Formula C8H13N3O4
Molecular Weight (g/mol) 215.209
MDL Number MFCD00149320
InChI Key PSWSDQRXCOJSFC-FJXQXJEOSA-N
Synonym ac-his-oh.h2o, n-acetyl-l-histidine hydrate, n-acetyl-l-histidine monohydrate, unii-pq2tc3x11o, s-2-acetamido-3-1h-imidazol-4-yl propanoic acid hydrate, pq2tc3x11o, acetyl histidine, 2s-2-acetylamino-3-1h-imidazol-4-yl propanoic acid hydrate, l-n-acetylhistidine monohydrate
PubChem CID 2724380
IUPAC Name (2S)-2-acetamido-3-(1H-imidazol-5-yl)propanoic acid;hydrate
SMILES CC(=O)NC(CC1=CN=CN1)C(=O)O.O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15457019
View Documents Promotion Details
Thermo Scientific Chemicals
J65657.03
1 g
48.90 €
1g
Please to purchase this item. Need a web account? Register with us today!
15467019
View Documents Promotion Details
Thermo Scientific Chemicals
J65657.06
5 g
159.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
15477019
View Documents Promotion Details
Thermo Scientific Chemicals
J65657.14
25 g
321.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

N-acetylhistidine, a novel osmolyte in the lens of Atlantic salmon. An ectotherm homologue of human predicted gene NAT16 encodes histidine N-acetyltransferase responsible for NÃŽ±-acetylhistidine synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-acetylhistidine, a novel osmolyte in the lens of Atlantic salmon. An ectotherm homologue of human predicted gene NAT16 encodes histidine N-acetyltransferase responsible for Nα-acetylhistidine synthesis.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

39145-52-3
215.209
PSWSDQRXCOJSFC-FJXQXJEOSA-N
2724380
CC(=O)NC(CC1=CN=CN1)C(=O)O.O
C8H13N3O4
MFCD00149320
ac-his-oh.h2o, n-acetyl-l-histidine hydrate, n-acetyl-l-histidine monohydrate, unii-pq2tc3x11o, s-2-acetamido-3-1h-imidazol-4-yl propanoic acid hydrate, pq2tc3x11o, acetyl histidine, 2s-2-acetylamino-3-1h-imidazol-4-yl propanoic acid hydrate, l-n-acetylhistidine monohydrate
(2S)-2-acetamido-3-(1H-imidazol-5-yl)propanoic acid;hydrate

Specifications

163°C
White
MFCD00149320
Soluble in water.
CC(=O)NC(CC1=CN=CN1)C(=O)O.O
215.209
215.21 (197.09 Anhydrous)
Powder
39145-52-3
C8H13N3O4
ac-his-oh.h2o, n-acetyl-l-histidine hydrate, n-acetyl-l-histidine monohydrate, unii-pq2tc3x11o, s-2-acetamido-3-1h-imidazol-4-yl propanoic acid hydrate, pq2tc3x11o, acetyl histidine, 2s-2-acetylamino-3-1h-imidazol-4-yl propanoic acid hydrate, l-n-acetylhistidine monohydrate
PSWSDQRXCOJSFC-FJXQXJEOSA-N
(2S)-2-acetamido-3-(1H-imidazol-5-yl)propanoic acid;hydrate
2724380
+47° (c=1 in Water)
N-Acetyl-L-histidine monohydrate
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only