missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-L-isoleucine, 98%, Thermo Scientific Chemicals
71.40 € - 388.00 €
Chemical Identifiers
CAS | 3077-46-1 |
---|---|
Molecular Formula | C8H15NO3 |
Molecular Weight (g/mol) | 173.21 |
MDL Number | MFCD00066072 |
InChI Key | JDTWZSUNGHMMJM-IFSCUNPJNA-N |
Synonym | n-acetyl-l-isoleucine, 2s,3s-2-acetamido-3-methylpentanoic acid, ac-ile-oh, l-isoleucine, n-acetyl, unii-p9cl0y0bvz, p9cl0y0bvz, isoleucine, n-acetyl-, l, acetyl isoleucine, n-acetyl-isoleucine, acetyl-l-isoleucine |
PubChem CID | 7036275 |
ChEBI | CHEBI:21555 |
SMILES | CC[C@H](C)[C@H](NC(C)=O)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15472819
|
Thermo Scientific Alfa Aesar
H66771.06 |
5 g |
71.40 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15482819
|
Thermo Scientific Alfa Aesar
H66771.14 |
25 g |
167.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15492819
|
Thermo Scientific Alfa Aesar
H66771.22 |
100 g | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3077-46-1 | |
173.21 | |
JDTWZSUNGHMMJM-IFSCUNPJNA-N | |
7036275 | |
CC[C@H](C)[C@H](NC(C)=O)C(O)=O |
C8H15NO3 | |
MFCD00066072 | |
n-acetyl-l-isoleucine, 2s,3s-2-acetamido-3-methylpentanoic acid, ac-ile-oh, l-isoleucine, n-acetyl, unii-p9cl0y0bvz, p9cl0y0bvz, isoleucine, n-acetyl-, l, acetyl isoleucine, n-acetyl-isoleucine, acetyl-l-isoleucine | |
CHEBI:21555 |
Specifications
3077-46-1 | |
C8H15NO3 | |
n-acetyl-l-isoleucine, 2s,3s-2-acetamido-3-methylpentanoic acid, ac-ile-oh, l-isoleucine, n-acetyl, unii-p9cl0y0bvz, p9cl0y0bvz, isoleucine, n-acetyl-, l, acetyl isoleucine, n-acetyl-isoleucine, acetyl-l-isoleucine | |
CC[C@H](C)[C@H](NC(C)=O)C(O)=O | |
173.21 | |
CHEBI:21555 | |
98% | |
N-Acetyl-L-isoleucine |
White | |
MFCD00066072 | |
JDTWZSUNGHMMJM-IFSCUNPJNA-N | |
(2S,3S)-2-acetamido-3-methylpentanoic acid | |
7036275 | |
173.21 | |
Crystalline |
Safety and Handling
missing translation for 'einecsNumber' : 221-364-9
missing translation for 'tsca' : No
missing translation for 'storageNote' : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only