missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-L-methionine, 99%, Thermo Scientific Chemicals
47.90 € - 415.00 €
Chemical Identifiers
CAS | 65-82-7 |
---|---|
Molecular Formula | C7H13NO3S |
Molecular Weight (g/mol) | 191.25 |
MDL Number | MFCD00064441 |
InChI Key | XUYPXLNMDZIRQH-LURJTMIESA-N |
Synonym | n-acetyl-l-methionine, n-acetylmethionine, acetyl-l-methionine, ac-met-oh, l-n-acetyl methionine, acetylmethionine, methionamine, methionine, n-acetyl-, l, thiomedon, l-methionine, n-acetyl |
PubChem CID | 448580 |
ChEBI | CHEBI:21557 |
IUPAC Name | (2S)-2-acetamido-4-methylsulfanylbutanoic acid |
SMILES | CSCC[C@H](NC(C)=O)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11445737
|
Thermo Scientific Alfa Aesar
B24312.06 |
5 g |
47.90 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11455737
|
Thermo Scientific Alfa Aesar
B24312.14 |
25 g |
141.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11465737
|
Thermo Scientific Alfa Aesar
B24312.22 |
100 g | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
65-82-7 | |
191.25 | |
XUYPXLNMDZIRQH-LURJTMIESA-N | |
448580 | |
(2S)-2-acetamido-4-methylsulfanylbutanoic acid |
C7H13NO3S | |
MFCD00064441 | |
n-acetyl-l-methionine, n-acetylmethionine, acetyl-l-methionine, ac-met-oh, l-n-acetyl methionine, acetylmethionine, methionamine, methionine, n-acetyl-, l, thiomedon, l-methionine, n-acetyl | |
CHEBI:21557 | |
CSCC[C@H](NC(C)=O)C(O)=O |
Specifications
103°C to 107°C | |
C7H13NO3S | |
1725552 | |
n-acetyl-l-methionine, n-acetylmethionine, acetyl-l-methionine, ac-met-oh, l-n-acetyl methionine, acetylmethionine, methionamine, methionine, n-acetyl-, l, thiomedon, l-methionine, n-acetyl | |
CSCC[C@H](NC(C)=O)C(O)=O | |
191.25 | |
CHEBI:21557 | |
99% | |
N-Acetyl-L-methionine |
65-82-7 | |
MFCD00064441 | |
14,96 | |
XUYPXLNMDZIRQH-LURJTMIESA-N | |
(2S)-2-acetamido-4-methylsulfanylbutanoic acid | |
448580 | |
191.25 | |
−21° (c=1.3 in Water) |
Safety and Handling
missing translation for 'einecsNumber' : 200-617-7
missing translation for 'rtecsNumber' : PD0480000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only