missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetylglycinamide, 97%, Thermo Scientific Chemicals
58.80 € - 704.00 €
Chemical Identifiers
CAS | 2620-63-5 |
---|---|
Molecular Formula | C4H8N2O2 |
Molecular Weight (g/mol) | 116.12 |
MDL Number | MFCD00008029 |
InChI Key | WQELDIQOHGAHEM-UHFFFAOYSA-N |
Synonym | n-acetylglycine amide, n-alpha-acetylglycinamide, unii-97sv9afw2z, acetamide, n-2-amino-2-oxoethyl, nalpha-acetylglycinamide, n-.alpha.-acetylglycinamide, 97sv9afw2z, ethanimidic acid, n-2-amino-2-oxoethyl, acetylglutamide, acetylglycinamide |
PubChem CID | 28326 |
IUPAC Name | 2-acetamidoacetamide |
SMILES | CC(=O)NCC(N)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11481457
|
Thermo Scientific Alfa Aesar
B24940.03 |
1 g |
58.80 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11491457
|
Thermo Scientific Alfa Aesar
B24940.06 |
5 g |
193.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11401467
|
Thermo Scientific Alfa Aesar
B24940.14 |
25 g |
704.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2620-63-5 | |
116.12 | |
WQELDIQOHGAHEM-UHFFFAOYSA-N | |
28326 | |
CC(=O)NCC(N)=O |
C4H8N2O2 | |
MFCD00008029 | |
n-acetylglycine amide, n-alpha-acetylglycinamide, unii-97sv9afw2z, acetamide, n-2-amino-2-oxoethyl, nalpha-acetylglycinamide, n-.alpha.-acetylglycinamide, 97sv9afw2z, ethanimidic acid, n-2-amino-2-oxoethyl, acetylglutamide, acetylglycinamide | |
2-acetamidoacetamide |
Specifications
2620-63-5 | |
Odorless | |
MFCD00008029 | |
n-acetylglycine amide, n-alpha-acetylglycinamide, unii-97sv9afw2z, acetamide, n-2-amino-2-oxoethyl, nalpha-acetylglycinamide, n-.alpha.-acetylglycinamide, 97sv9afw2z, ethanimidic acid, n-2-amino-2-oxoethyl, acetylglutamide, acetylglycinamide | |
CC(=O)NCC(N)=O | |
116.12 | |
116.12 | |
N-Acetylglycinamide |
136°C to 140°C | |
C4H8N2O2 | |
1753982 | |
WQELDIQOHGAHEM-UHFFFAOYSA-N | |
2-acetamidoacetamide | |
28326 | |
97% |
Safety and Handling
missing translation for 'einecsNumber' : 220-058-2
missing translation for 'rtecsNumber' : MB7507000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only