Learn More
Thermo Scientific Chemicals N(alpha)-Acetyl-L-lysine, 99%
71.20 € - 263.00 €
Chemical Identifiers
CAS | 1946-82-3 |
---|---|
Molecular Formula | C8H16N2O3 |
Molecular Weight (g/mol) | 188.23 |
MDL Number | MFCD00008233 |
InChI Key | VEYYWZRYIYDQJM-JLDDOWRYNA-N |
Synonym | ac-lys-oh, n-alpha-acetyl-l-lysine, n alpha-acetyllysine, nalpha-acetyl-l-lysine, n-acetyl-l-lysine, n2-acetyl-l-lysine, s-2-acetamido-6-aminohexanoic acid, n alpha-acetyl-l-lysine, n 2-acetyl-l-lysine, acetyl-l-lysine |
PubChem CID | 92907 |
ChEBI | CHEBI:35704 |
IUPAC Name | (2S)-2-acetamido-6-aminohexanoic acid |
SMILES | CC(=O)N[C@@H](CCCCN)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15406719
|
Thermo Scientific Chemicals
J65083.03 |
1 g |
71.20 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15416719
|
Thermo Scientific Chemicals
J65083.06 |
5 g |
263.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nα-Acetyl-L-lysine is a biologically available N-terminal capped form of the proteinogenic α amino acid L-lysine. Nα-Acetyl-L-lysine is a substrate used to study and characterize lysyl oxidize(s).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsNα-Acetyl-L-lysine is a biologically available N-terminal capped form of the proteinogenic α amino acid L-lysine. Nα-Acetyl-L-lysine is a substrate used to study and characterize lysyl oxidize(s).
Solubility
Soluble in water (miscible), and 80% acetic acid (50 mg/ml--clear colorless solution)).
Notes
Protect from heat. Store away from oxidizing agents.
Chemical Identifiers
1946-82-3 | |
188.23 | |
VEYYWZRYIYDQJM-JLDDOWRYNA-N | |
92907 | |
(2S)-2-acetamido-6-aminohexanoic acid |
C8H16N2O3 | |
MFCD00008233 | |
ac-lys-oh, n-alpha-acetyl-l-lysine, n alpha-acetyllysine, nalpha-acetyl-l-lysine, n-acetyl-l-lysine, n2-acetyl-l-lysine, s-2-acetamido-6-aminohexanoic acid, n alpha-acetyl-l-lysine, n 2-acetyl-l-lysine, acetyl-l-lysine | |
CHEBI:35704 | |
CC(=O)N[C@@H](CCCCN)C(O)=O |
Specifications
256°C to 258°C (decomposition) | |
White | |
MFCD00008233 | |
Soluble in water (miscible),and 80% acetic acid (50mg/ml--clear colorless solution)). | |
CC(=O)N[C@@H](CCCCN)C(O)=O | |
188.23 | |
CHEBI:35704 | |
99% | |
N(alpha)-Acetyl-L-lysine |
1946-82-3 | |
C8H16N2O3 | |
ac-lys-oh, n-alpha-acetyl-l-lysine, n alpha-acetyllysine, nalpha-acetyl-l-lysine, n-acetyl-l-lysine, n2-acetyl-l-lysine, s-2-acetamido-6-aminohexanoic acid, n alpha-acetyl-l-lysine, n 2-acetyl-l-lysine, acetyl-l-lysine | |
VEYYWZRYIYDQJM-JLDDOWRYNA-N | |
(2S)-2-acetamido-6-aminohexanoic acid | |
92907 | |
188.2 | |
Powder |
Safety and Handling
missing translation for 'einecsNumber' : 217-747-5
missing translation for 'tsca' : No
Recommended Storage : Store at -20°C
RUO – Research Use Only