missing translation for 'onlineSavingsMsg'
Learn More

N(alpha)-Benzyloxycarbonyl-D-histidine, 98+%, Thermo Scientific™

Product Code. 11372826
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67424-93-5
Molecular Formula C14H15N3O4
Molecular Weight (g/mol) 289.291
MDL Number MFCD00067702
InChI Key WCOJOHPAKJFUDF-GFCCVEGCSA-N
Synonym z-d-his-oh, nalpha-cbz-d-histidine, cbz-d-his-oh, cbz-d-histidine, z-d-histidine, nalpha-carbobenzoxy-d-histidine, n alpha-cbz-d-histidine, n alpha-benzyloxycarbonyl-d-histidine, n-benzyloxy carbonyl-d-histidine, d-histidine,n-phenylmethoxy carbonyl
PubChem CID 736161
IUPAC Name (2R)-3-(1H-imidazol-5-yl)-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CC2=CN=CN2)C(=O)O

Specifications

Melting Point 161°C to 163°C
Beilstein 92342
Formula Weight 289.29
Percent Purity ≥98%
Quantity 1 g
Optical Rotation +24° (c=6 in 6N HCl)
Chemical Name or Material N(alpha)-Benzyloxycarbonyl-D-histidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'