missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzoyl-2'-deoxyadenosine, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
Reactant in the synthesis of 2', 5'-Dideoxycytidines and Other Derivatives of 2'-Deoxycytidine. Reactant in the synthesis of Mono- and Diamino Analogues of 2?-Deoxyadenosine, Cordycepin, 9-(3-Deoxy-α-D-Threo-Pentofuranosyl)-Adenine and 9-(2-Deoxy-α-D-Threo-Pentofuranosyl)Adenine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Reactant in the synthesis of 2′, 5′-Dideoxycytidines and Other Derivatives of 2′-Deoxycytidine. Reactant in the synthesis of Mono- and Diamino Analogues of 2′-Deoxyadenosine, Cordycepin, 9-(3-Deoxy-α-D-Threo-Pentofuranosyl)-Adenine and 9-(2-Deoxy-α-D-Threo-Pentofuranosyl)Adenine.
Solubility
Soluble in water (slightly), and methanol (20 mg/ml).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Reactant in the synthesis of 2′, 5′-Dideoxycytidines and Other Derivatives of 2′-Deoxycytidine. Reactant in the synthesis of Mono- and Diamino Analogues of 2′-Deoxyadenosine, Cordycepin, 9-(3-Deoxy-α-D-Threo-Pentofuranosyl)-Adenine and 9-(2-Deoxy-α-D-Threo-Pentofuranosyl)Adenine.
Solubility
Soluble in water (slightly), and methanol (20 mg/ml).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
Quantity | 250 mg |
Synonym | n6-benzoyl-2'-deoxyadenosine, n-benzoyl-2'-deoxyadenosine, n-benzoyl-2'-deoxy-adenosine, n-9-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-9h-purin-6-yl benzamide, n 6-benzoyl-2'-deoxyadenosine, n6-benzoyl-2-deoxyadenosine, adenosine, n-benzoyl-2'-deoxy, n-benzoyldeoxyadenosine, n6-benzoyldeoxyadenosine, 6-n-benzoyldeoxyadenosine |
Solubility Information | Soluble in water (slightly),and methanol (20mg/ml). |
Formula Weight | 355.35 |
Percent Purity | ≥98% |
Chemical Name or Material | N-Benzoyl-2'-deoxyadenosine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'