missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzyloxycarbonyl-L-glutamic acid 1-methyl ester, 98%, Thermo Scientific Chemicals
102.00 € - 374.00 €
Chemical Identifiers
CAS | 5672-83-3 |
---|---|
Molecular Formula | C14H17NO6 |
Molecular Weight (g/mol) | 295.291 |
MDL Number | MFCD00083278 |
InChI Key | BGMCTGARFXPQML-NSHDSACASA-N |
Synonym | z-glu-ome, z-l-glu-ome, 1-methyl n-carbobenzoxy-l-glutamate, 4s-4-benzyloxy carbonyl amino-5-methoxy-5-oxopentanoic acid, cbz-glu-ome, l-glutamic acid,n-phenylmethoxy carbonyl-, 1-methyl ester, n-cbz-l-glu-ome, z-l-glutamic acid 1-methyl ester, n-cbz-l-glutamic acid 1-methyl ester, n-carbobenzoxy-l-glutamic acid 1-methyl ester |
PubChem CID | 2733408 |
IUPAC Name | (4S)-5-methoxy-5-oxo-4-(phenylmethoxycarbonylamino)pentanoic acid |
SMILES | COC(=O)C(CCC(=O)O)NC(=O)OCC1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15440646
|
Thermo Scientific Alfa Aesar
H53417.03 |
1 g |
102.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15450646
|
Thermo Scientific Alfa Aesar
H53417.06 |
5 g |
374.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5672-83-3 | |
295.291 | |
BGMCTGARFXPQML-NSHDSACASA-N | |
2733408 | |
COC(=O)C(CCC(=O)O)NC(=O)OCC1=CC=CC=C1 |
C14H17NO6 | |
MFCD00083278 | |
z-glu-ome, z-l-glu-ome, 1-methyl n-carbobenzoxy-l-glutamate, 4s-4-benzyloxy carbonyl amino-5-methoxy-5-oxopentanoic acid, cbz-glu-ome, l-glutamic acid,n-phenylmethoxy carbonyl-, 1-methyl ester, n-cbz-l-glu-ome, z-l-glutamic acid 1-methyl ester, n-cbz-l-glutamic acid 1-methyl ester, n-carbobenzoxy-l-glutamic acid 1-methyl ester | |
(4S)-5-methoxy-5-oxo-4-(phenylmethoxycarbonylamino)pentanoic acid |
Specifications
5672-83-3 | |
C14H17NO6 | |
MFCD00083278 | |
z-glu-ome, z-l-glu-ome, 1-methyl n-carbobenzoxy-l-glutamate, 4s-4-benzyloxy carbonyl amino-5-methoxy-5-oxopentanoic acid, cbz-glu-ome, l-glutamic acid,n-phenylmethoxy carbonyl-, 1-methyl ester, n-cbz-l-glu-ome, z-l-glutamic acid 1-methyl ester, n-cbz-l-glutamic acid 1-methyl ester, n-carbobenzoxy-l-glutamic acid 1-methyl ester | |
−24° (c=1 in Ethanol) | |
(4S)-5-methoxy-5-oxo-4-(phenylmethoxycarbonylamino)pentanoic acid | |
2733408 | |
98% |
64°C to 70°C | |
1.4365 | |
2567599 | |
BGMCTGARFXPQML-NSHDSACASA-N | |
COC(=O)C(CCC(=O)O)NC(=O)OCC1=CC=CC=C1 | |
295.291 | |
295.29 | |
N-Benzyloxycarbonyl-L-glutamic acid 1-methyl ester |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only