missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-alanine, 98+%, Thermo Scientific Chemicals

42.00 € - 148.00 €

Chemical Identifiers

CAS 7764-95-6
Molecular Formula C8H15NO4
Molecular Weight (g/mol) 189.21
MDL Number MFCD00063123
InChI Key QVHJQCGUWFKTSE-RXMQYKEDSA-N
Synonym boc-d-alanine, boc-d-ala-oh, n-boc-d-alanine, n-tert-butoxycarbonyl-d-alanine, r-2-tert-butoxycarbonyl amino propanoic acid, n-alpha-t-boc-d-alanine, boc-d-ala, n-t-butoxycarbonyl-d-alanine, 2r-2-tert-butoxy carbonyl amino propanoic acid
PubChem CID 637606
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES C[C@@H](NC(=O)OC(C)(C)C)C(O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11425357
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B22706.06
5 g
42.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
11435357
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B22706.14
25 g
148.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7764-95-6
189.21
QVHJQCGUWFKTSE-RXMQYKEDSA-N
637606
C[C@@H](NC(=O)OC(C)(C)C)C(O)=O
C8H15NO4
MFCD00063123
boc-d-alanine, boc-d-ala-oh, n-boc-d-alanine, n-tert-butoxycarbonyl-d-alanine, r-2-tert-butoxycarbonyl amino propanoic acid, n-alpha-t-boc-d-alanine, boc-d-ala, n-t-butoxycarbonyl-d-alanine, 2r-2-tert-butoxy carbonyl amino propanoic acid
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid

Specifications

7764-95-6
C8H15NO4
2048396
QVHJQCGUWFKTSE-RXMQYKEDSA-N
C[C@@H](NC(=O)OC(C)(C)C)C(O)=O
189.21
189.21
N-Boc-D-alanine
81°C to 84°C
MFCD00063123
boc-d-alanine, boc-d-ala-oh, n-boc-d-alanine, n-tert-butoxycarbonyl-d-alanine, r-2-tert-butoxycarbonyl amino propanoic acid, n-alpha-t-boc-d-alanine, boc-d-ala, n-t-butoxycarbonyl-d-alanine, 2r-2-tert-butoxy carbonyl amino propanoic acid
+26° (c=2 in Acetic Acid)
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
637606
≥98%
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only