Learn More
N-Boc-D-phenylalanine methyl ester, 98%, Thermo Scientific Chemicals
Description
N-Boc-D-phenylalanine methyl ester employed as a primary and secondary intermediate chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
N-Boc-D-phenylalanine methyl ester employed as a primary and secondary intermediate chemical research.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Specifications
Specifications
Solubility Information | Slightly soluble in water. |
IUPAC Name | methyl 2-{[(tert-butoxy)carbonyl]amino}-3-phenylpropanoate |
Quantity | 25 g |
Formula Weight | 279.34 |
Percent Purity | 98% |
Physical Form | Solid |
Chemical Name or Material | N-Boc-D-phenylalanine methyl ester |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.