missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-aspartic acid, 98%, Thermo Scientific Chemicals

Product Code. 11358195
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13726-67-5
Molecular Formula C9H15NO6
Molecular Weight (g/mol) 233.22
MDL Number MFCD00037279
InChI Key KAJBMCZQVSQJDE-YFKPBYRVSA-N
Synonym boc-asp-oh, n-tert-butoxycarbonyl-l-aspartic acid, boc-l-aspartic acid, boc-l-asp-oh, n-boc-l-aspartic acid, boc-aspartic acid, boc-l-aspartic acid-oh, n-tert-butyloxycarbonyl-l-aspartic acid, n-t-butoxycarbonyl-l-aspatic acid, 2s-2-tert-butoxycarbonyl amino butanedioic acid
PubChem CID 99718
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanedioic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)O)C(=O)O

Specifications

Melting Point 115°C
Quantity 1 g
Beilstein 1913973
Optical Rotation −5.5° (c=1 in Methanol)
Formula Weight 233.23
Percent Purity 98%
Chemical Name or Material N-Boc-L-aspartic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'