missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-trans-4-hydroxy-L-proline, 97%, Thermo Scientific Chemicals

Product Code. 11466200
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13726-69-7
Molecular Formula C10H17NO5
Molecular Weight (g/mol) 231.25
MDL Number MFCD00053370
InChI Key BENKAPCDIOILGV-UHFFFAOYNA-N
Synonym boc-hyp-oh, boc-l-hydroxyproline, 2s,4r-1-tert-butoxycarbonyl-4-hydroxypyrrolidine-2-carboxylic acid, boc-trans-4-hydroxy-l-proline, n-boc-trans-4-hydroxy-l-proline, boc-l-hyp-oh, boc-hydroxyproline, unii-rc99p5b8ig, trans-boc-hyp-oh, 4r-1-tert-butoxycarbonyl-4-hydroxy-l-proline
PubChem CID 88804
IUPAC Name (2S,4R)-4-hydroxy-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CC(O)CC1C(O)=O

Specifications

Melting Point 123°C to 127°C
Quantity 1 g
Formula Weight 231.25
Percent Purity 97%
Chemical Name or Material N-Boc-trans-4-hydroxy-L-proline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'