missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-D-aspartic acid 1-tert-butyl ester, 95%, Thermo Scientific Chemicals

Product Code. 15417726
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

N-Fmoc-D-aspartic acid 1-tert-butyl ester is an pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-D-aspartic acid 1-tert-butyl ester is an pharmaceutical intermediate.

Solubility
Soluble in DMF.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134098-70-7
Molecular Formula C23H25NO6
Molecular Weight (g/mol) 411.454
MDL Number MFCD00237011
InChI Key VZXQYACYLGRQJU-LJQANCHMSA-N
Synonym fmoc-d-asp-otbu, fmoc-d-aspartic acid 1-tert-butyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-d-aspartic acid alpha-t-butyl ester, 3r-4-tert-butoxy-3-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxobutanoic acid, r-3-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxy-4-oxobutanoic acid, fmoc-d-asp otbu, ambotzfaa1311, fmoc-d-aspartic acid-otbu, fmoc-d-asp-tert-butoxide, fmoc-d-aspartic acid alpha-t-butyl ester
PubChem CID 7019703
IUPAC Name (3R)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid
SMILES CC(C)(C)OC(=O)C(CC(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Quantity 250 mg
Solubility Information Soluble in DMF.
Formula Weight 411.45
Percent Purity 95%
Chemical Name or Material N-Fmoc-D-aspartic acid 1-tert-butyl ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'