missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-aspartic acid, 95%, Thermo Scientific Chemicals

Product Code. 15488818
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

N-Fmoc-L-aspartic acid is used to biosynthesize other amino acids with in the human body.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-L-aspartic acid is used to biosynthesize other amino acids with in the human body.

Solubility
Soluble in methanol, and water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119062-05-4
Molecular Formula C19H17NO6
Molecular Weight (g/mol) 355.346
MDL Number MFCD00237654
InChI Key KSDTXRUIZMTBNV-INIZCTEOSA-N
Synonym fmoc-asp-oh, fmoc-l-aspartic acid, n-fmoc-l-aspartic acid, fmoc-l-asp-oh, fmoc-aspartic acid, n-9h-fluoren-9-ylmethoxy carbonyl-l-aspartic acid, s-2-9h-fluoren-9-yl methoxy carbonylamino succinic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino succinic acid, aspartic acid, n-9h-fluoren-9-ylmethoxy carbonyl, n-9-fluorenylmethoxycarbonyl-l-aspartic acid
PubChem CID 7019016
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)butanedioic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC(=O)O)C(=O)O

Specifications

Quantity 100 g
Beilstein 3632557
Solubility Information Soluble in methanol,and water.
Formula Weight 355.34
Percent Purity 95%
Chemical Name or Material N-Fmoc-L-aspartic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'