missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-glutamic acid 5-tert-butyl ester, 98%, Thermo Scientific Chemicals

Product Code. 15405408
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71989-18-9
Molecular Formula C24H27NO6
Molecular Weight (g/mol) 425.481
MDL Number MFCD00037135
InChI Key OTKXCALUHMPIGM-FQEVSTJZSA-N
Synonym fmoc-glu otbu-oh, fmoc-l-glutamic acid 5-tert-butyl ester, 2s-5-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-oxopentanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid, n-fmoc-l-glutamic acid 5-tert-butyl ester, fmoc-glu obut, fmoc-glu tbu, fmoc-l-glu otbu, pubchem12950, fmoc-l-glu otbu-oh
PubChem CID 2724637
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid
SMILES CC(C)(C)OC(=O)CCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Melting Point 83°C to 90°C
Quantity 25 g
Beilstein 3636375
Optical Rotation −5.0° (c=1 in Acetic Acid/Water (4:1))
Formula Weight 425.48
Percent Purity 98%
Chemical Name or Material N-Fmoc-L-glutamic acid 5-tert-butyl ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'