missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-N-methyl-D-phenylalanine, 95%

Product Code. 15409528
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15409528

Brand: Thermo Scientific Alfa Aesar H63055.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 138775-05-0 | C25H23NO4 | 401.462 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138775-05-0
Molecular Formula C25H23NO4
Molecular Weight (g/mol) 401.462
MDL Number MFCD00153392
InChI Key GBROUWPNYVBLFO-HSZRJFAPSA-N
Synonym fmoc-n-methyl-d-phenylalanine, fmoc-d-mephe-oh, fmoc-n-me-d-phe-oh, r-2-9h-fluoren-9-yl methoxy carbonyl methyl amino-3-phenylpropanoic acid, fmoc-nalpha-methyl-d-phenylalanine, d-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, 2r-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino-3-phenylpropanoic acid, ambotzfaa1333, fmoc-nme d phe-oh, fmoc-n-, a-methyl-d-phenylalanine
PubChem CID 978357
IUPAC Name (2R)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-3-phenylpropanoic acid
SMILES CN(C(CC1=CC=CC=C1)C(=O)O)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Beilstein 8441627
Quantity 5 g
Formula Weight 401.45
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Fmoc-N-methyl-D-phenylalanine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.