missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-N-methyl-L-leucine, 98%, Thermo Scientific Chemicals

Product Code. 15410159
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103478-62-2
Molecular Formula C22H25NO4
Molecular Weight (g/mol) 367.45
MDL Number MFCD00151933
InChI Key BUJQSIPFDWLNDC-GNLPSFAGNA-N
Synonym fmoc-n-methyl-l-leucine, fmoc-n-me-leu-oh, n-fmoc-n-methyl-l-leucine, fmoc-meleu-oh, l-leucine, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, 2s-2-9h-fluoren-9-ylmethoxycarbonyl methyl amino-4-methyl-pentanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino-4-methylpentanoic acid, pubchem13155, fmoc-n-a-methyl-l-leucine, n-methyl-n-fmoc-l-leu-oh
PubChem CID 7015835
SMILES CC(C)C[C@H](N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 113°C to 116°C
Color White
Beilstein 3630430
Solubility Information Slightly soluble in water.
IUPAC Name (2S)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-4-methylpentanoic acid
Formula Weight 367.44
Percent Purity 98%
Quantity 250 mg
Physical Form Powder
Chemical Name or Material N-Fmoc-N-methyl-L-leucine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'