missing translation for 'onlineSavingsMsg'
Learn More

N-Methylpyrrolidinone, Fisher BioReagents

Product Code. p-27176
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
Unit Size:
4 L
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 10317033

Brand: Fisher BioReagents BP11724

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

For automated oligonucleotide synthesis

N-Methylpyrrolidinone is used in automated oligonucleotide synthesis.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 872-50-4
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.133
InChI Key SECXISVLQFMRJM-UHFFFAOYSA-N
Synonym 1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl
PubChem CID 13387
ChEBI CHEBI:7307
IUPAC Name 1-methylpyrrolidin-2-one
SMILES CN1CCCC1=O

Specifications

pH 7.7 to 8.0
Melting Point -24°C
Absorbance 0.02 AU max. at 400nm, 0.3 AU max. at 300nm, 0.7 AU max. at 270nm, 1.0 AU max. at 265nm
Color Colorless
Density 1.030g/cm³
Boiling Point 202°C
Quantity 4 L
Packaging Glass Bottle
Physical Form Liquid
Assay Percent Range (by GC): ≥99.8%
Titratable Base 0.001mEq/g max.
Vapor Pressure 0.7mbar at 25°C
Water 0.05% max.
Merck Index 15, 6190
Formula Weight 99.13
Percent Purity ≥99.8%
Chemical Name or Material N-Methylpyrrolidinone
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.