missing translation for 'onlineSavingsMsg'
Learn More

N6-(4-Hydroxybenzyl)adenosine, MedChemExpress

Product Code. 16413647
Encompass_Preferred
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
50 mg
100 mg
missing translation for 'unitSize'
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16413647

missing translation for 'mfr': Medchem Express HY18775

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress N6-(4-Hydroxybenzyl)adenosine is a inhibitor of platelet aggregation induced in vitro by collagen and their activity range was demonstrated (IC50: 6.77-141 μM).

N6-(4-Hydroxybenzyl)adenosine is a inhibitor of platelet aggregation induced in vitro by collagen and their activity range was demonstrated (IC50: 6.77-141 μM).

Specifications

Chemical Name or Material N6-(4-Hydroxybenzyl)adenosine
CAS 110505-75-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C17H19N5O5
Quantity 50 mg
Synonym Para-topolin riboside
Solubility Information DMSO : ≥ 100 mg/mL (267.84 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES OC[C@@H]1[C@H]([C@H]([C@H](N2C=NC3=C2N=CN=C3NCC4=CC=C(O)C=C4)O1)O)O
Molecular Weight (g/mol) 373.36
Formula Weight 373.36
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.