Learn More
Macherey-Nagel™ N9 Polypropylene Green Screw Cap
Product Code. 12735232 Shop All Macherey-Nagel Products
Click to view available options
Color:
Green
Unit Size:
Pack of 100
This item is not returnable. View return policy
Product Code. 12735232
Brand: Macherey-Nagel™ 702083
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Color Green
Material Polypropylene, PTFE, Silicone
Septum Silicone white/PTFE blue, slit
Thickness (Metric) 1 mm
Closure Size N9