missing translation for 'onlineSavingsMsg'
Learn More

NAG Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP3-18441V

Product Code. 18594918

  • 663.00 € / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

NAG Polyclonal antibody specifically detects NAG in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NAG
Polyclonal
Western Blot
DKFZp586G1219, EC 1.1.1.94, EC 1.2.1.59, FLJ40407, NAG/BC035112 fusion, NAG/FAM49A fusion, NAGneuroblastoma-amplified protein, neuroblastoma amplified sequence, Neuroblastoma-amplified gene protein, neuroblastoma-amplified sequence
Recognizes region between residues Between Between 350 and 400..
0.1 mL
Cell Biology
51594
Store at 4°C in the dark.
IgG
Western Blot
DyLight 405
50mM Sodium Borate
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
NAG Antibody, DyLight 405, Novus Biologicals™ > 0.1 mL; DyLight 405

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.