Learn More
Invitrogen™ Naked1 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5113280
Product Code. 17111144
  • 457.00 € / 100µg
Please to purchase this item. Need a web account? Register with us today!
Description
Description
Naked1 Polyclonal Antibody for Western Blot, ICC/IF, ELISA
In the mouse, Nkd is a Dishevelled (see DVL1; MIM 601365)-binding protein that functions as a negative regulator of the Wnt (see WNT1; MIM 164820)-beta-catenin-Tcf signaling pathway.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Naked1
Polyclonal
Unconjugated
NKD1
2810434J10Rik; 9030215G15Rik; Dvl-binding protein; hNkd; hNkd1; mNkd; mNkd1; naked cuticle 1 homolog; naked cuticle 1 homolog (Drosophila); naked cuticle homolog 1; naked cuticle homolog 1 (Drosophila); naked cuticle-1; Naked1; naked-1; Nkd; NKD1; PP7246; Protein naked cuticle homolog 1
Rabbit
Protein G
RUO
364952, 85407
-20°C or -80°C if preferred
Liquid
ELISA, Western Blot, Immunocytochemistry
3.52 mg/mL
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Q969G9
NKD1
Recombinant Human Protein naked cuticle homolog 1 protein (269-384AA).
100 μg
Primary
Human, Rat