Learn More
Thermo Scientific™ Nalgene™ Staining Box
GREENER_CHOICE
Product Code. p-4528941
Click to view available options
Type:
Gel Staining Box
Staining Box Plug
Unit Size:
Each
Pack of 12
Product Code. 10492491
Brand: Thermo Scientific™ 57051010
Please to purchase this item. Need a web account? Register with us today!
Edge
Stain, fix and handle fragile electrophoresis gels and membranes with less chance of damage than conventional methods with these staining boxes.
Stain, fix and handle fragile electrophoresis gels and membranes with less chance of damage than conventional methods. Thermo Scientific™ Nalgene™ Staining Box is equipped with a bottom drain allowing you to change solution without disturbing the box's other fragile contents. Box holds 4.8 in. x 4.8 in (189 mm x 189 mm) gel along with 500mL of liquid.
  • Polymethylpentene box and polypropylene copolymer cover
  • Useful over a -70° to +70°C (-94° to +158°F) temperature range
  • Good chemical compatibility with acids, bases and select organic solvents for a versatile range of protocols
  • LDPE plug in bottom for easy drainage of liquid with minimal handling of gel and provides a leakproof seal
  • Plug won't interfere with cover or contents of boxes on a benchtop or shaker
  • Stackable, with a handle on each end for safe, easy carrying
  • Microwavable and easy to clean
TRUSTED_SUSTAINABILITY
Specifications
Type Gel Staining Box
Material PMP
No. per Case 2
No. per Pack 1
Dimensions (L x W x H) 5 x 5 x 2 in. (12.7 x 12.7 x 5.08 cm)
Length (English) 5 in.
Length (Metric) 12.7 cm
Width (English) 5 in.
Width (Metric) 12.7 cm
Height (English) 2 in.
Height (Metric) 5.08 cm