Learn More
Reitenspiess™ Test Tube Brush with Wool Tip
missing translation for 'orderingAttributeHoverText'
Length (Metric):
220 mm
260 mm
270 mm
280 mm
missing translation for 'unitSize'
Each
Description
- Natural bristles
- Wool Tip
- Sterile
Specifications
Specifications
| Product Type | Test Tube Brush |
| Sterility | Sterile |
| For Use With (Application) | For cleaning, disinfection, sterilization. |
| Length (Metric) | 280 mm |
| Quantity | 1 Ea. |