Learn More
Memmert™ Nearly Gastight Interior with Gas Inlet/Outlet for UF1060
Nearly gastight interior with possibility for inlet/outlet through two tubes with ball valves.
Brand: Memmert™ UF1060K3
Description
- Gas inlet/outlet through 2 tubes with ball valves
- Technical clarification required for combination with other options
Specifications
UF1060 Oven | |
Interior | |
Gas Inlet/Outlet |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.