Learn More
Netilmicin sulfate, MedChemExpress
MedChemExpress Netilmicin (sulfate) (SCH-20569 (sulfate)) is an active aminoglycoside antibiotic against most Gram-negative and some Gram-positive bacteria, including certain strains resistant to gentamicin.
Brand: Medchem Express HY-A0086
This item is not returnable.
View return policy
Description
Netilmicin (sulfate) (SCH-20569 (sulfate)) is an active aminoglycoside antibiotic against most Gram-negative and some Gram-positive bacteria, including certain strains resistant to gentamicin.Specifications
Netilmicin sulfate | |
Yellow | |
C21H41N5O7.5/2H2SO4 | |
SCH-20569 sulfate | |
NCC1=CC[C@@H](N)[C@](O[C@@H]2[C@@H](N)C[C@@H](NCC)[C@H](O[C@@]3([H])OC[C@](C)(O)[C@H](NC)[C@H]3O)[C@H]2O)([H])O1.[2.5H2SO4] | |
720.78 | |
Research | |
COVID-19-immunoregulation |
56391-57-2 | |
Research | |
100 mg | |
H2O : 100 mg/mL (138.74 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (1.39 mM; ultrasonic and warming and heat to 80°C) | |
720.78 | |
98.0% | |
Solid |