Learn More
Netilmicin sulfate, MedChemExpress
MedChemExpress Netilmicin (sulfate) (SCH-20569 (sulfate)) is an active aminoglycoside antibiotic against most Gram-negative and some Gram-positive bacteria, including certain strains resistant to gentamicin.
Brand:  Medchem Express HY-A0086
Encompass_Preferred
 View more versions of this product
Product Code. 16460728
  • 87.75 € / 5mg
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
This item is not returnable. View return policy
Description
Description
Netilmicin (sulfate) (SCH-20569 (sulfate)) is an active aminoglycoside antibiotic against most Gram-negative and some Gram-positive bacteria, including certain strains resistant to gentamicin.
Specifications
Specifications
Netilmicin sulfate
Yellow
C21H41N5O7.5/2H2SO4
SCH-20569 sulfate
NCC1=CC[C@@H](N)[C@](O[C@@H]2[C@@H](N)C[C@@H](NCC)[C@H](O[C@@]3([H])OC[C@](C)(O)[C@H](NC)[C@H]3O)[C@H]2O)([H])O1.[2.5H2SO4]
720.78
Research
COVID-19-immunoregulation
56391-57-2
Research
5 mg
H2O : 100 mg/mL (138.74 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (1.39 mM; ultrasonic and warming and heat to 80°C)
720.78
98.0%
Solid