missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals NG-Monomethyl-L-arginine monoacetate, 99%

44.50 € - 398.00 €

Chemical Identifiers

CAS 53308-83-1
Molecular Formula C9H20N4O4
Molecular Weight (g/mol) 248.283
MDL Number MFCD00069311
InChI Key IKPNWIGTWUZCKM-JEDNCBNOSA-N
Synonym tilarginine acetate, l-nmma, l-nmma acetate, ng-monomethyl-l-arginine acetate, nomega-me-l-arg, unii-2fl3530af2, ng-me-l-arg, acoh, tilarginine acetate usan, ng-methyl-l-arginine acetate salt, l-ornithine, n5-imino methylamino methyl-cas
PubChem CID 135242
IUPAC Name acetic acid;(2S)-2-amino-5-[(N'-methylcarbamimidoyl)amino]pentanoic acid
SMILES CC(=O)O.CN=C(N)NCCCC(C(=O)O)N
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10173101
View Documents Promotion Details
Thermo Scientific Chemicals
328720050
5 mg Glass Bottle
44.50 €
5mg
Please to purchase this item. Need a web account? Register with us today!
10016213
View Documents Promotion Details
Thermo Scientific Chemicals
328720250
25 mg Glass Bottle
148.00 €
25mg
Please to purchase this item. Need a web account? Register with us today!
10371484
View Documents Promotion Details
Thermo Scientific Chemicals
328721000
100 mg Glass Bottle
398.00 €
100mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

53308-83-1
248.283
IKPNWIGTWUZCKM-JEDNCBNOSA-N
135242
CC(=O)O.CN=C(N)NCCCC(C(=O)O)N
C9H20N4O4
MFCD00069311
tilarginine acetate, l-nmma, l-nmma acetate, ng-monomethyl-l-arginine acetate, nomega-me-l-arg, unii-2fl3530af2, ng-me-l-arg, acoh, tilarginine acetate usan, ng-methyl-l-arginine acetate salt, l-ornithine, n5-imino methylamino methyl-cas
acetic acid;(2S)-2-amino-5-[(N'-methylcarbamimidoyl)amino]pentanoic acid

Specifications

180°C to 190°C
98.5
White
C9H20N4O4
MFCD00069311
tilarginine acetate, l-nmma, l-nmma acetate, ng-monomethyl-l-arginine acetate, nomega-me-l-arg, unii-2fl3530af2, ng-me-l-arg, acoh, tilarginine acetate usan, ng-methyl-l-arginine acetate salt, l-ornithine, n5-imino methylamino methyl-cas
CC(=O)O.CN=C(N)NCCCC(C(=O)O)N
+9° to +13° (20°C, 589nm) (c=4, water)
135242
≥98.5%
Powder
53308-83-1
100.0
99%
Authentic
15, 6096
IKPNWIGTWUZCKM-JEDNCBNOSA-N
acetic acid;(2S)-2-amino-5-[(N'-methylcarbamimidoyl)amino]pentanoic acid
248.283
248.3
+ 10.50 (20.00°C c=4,water)
N-ω-Monomethyl-L-arginine monoacetate
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only