Learn More
R&D Systems NHP XL CCL2/MCP-1, Magnetic Luminex™ Performance Assay
Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.
Brand:  R&D Systems LNHPXL279
Product Code. 18774524
  • 227.00 € / 1 set
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
This item is not returnable. View return policy
Description
Description
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 Kit
CCL2/JE/MCP-1
C-C motif chemokine ligand 2, CCL2, GDCF-2, HC11, HSMCR30, MCAF, Mcp1, MCP-1, SCYA2, SMC-CF
Primate
6347
Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&D Systems NHP XL CCL2/MCP-1, Magnetic Luminex™ Performance Assay > 1 Kit