Learn More
Reichert™ NIST Certified Calibration Oil
Check the accuracy of an instrument using calibration oils. Reichert™ NIST Certified Calibration Oil are formulated laser liquid calibration oils that are supplied with an NIST certification of traceability.
Brand:  Reichert™ 13K41390
 View more versions of this product
Product Code. 16317651
  • 441.00 € / Each
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
Description
Description
Formulated, measured, and certified to five significant places of refractive index (0.00001).
Specifications
Specifications
NIST Certified Calibration Oil
1.35999
NIST certification of traceability
18.04 Brix
Refractometer
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Reichert™ NIST Certified Calibration Oil > 1.36000 RI; Approximate Brix Value: 18.04