Learn More
Reichert™ NIST Certified Calibration Oil
Product Code. p-7079207
Click to view available options
Refractive Index:
1.35999
Scale Range:
18.04 Brix
Unit Size:
Each
Product Code. 16317651
Brand: Reichert™ 13K41390
Please to purchase this item. Need a web account? Register with us today!
Check the accuracy of an instrument using calibration oils. Reichert™ NIST Certified Calibration Oil are formulated laser liquid calibration oils that are supplied with an NIST certification of traceability.
Formulated, measured, and certified to five significant places of refractive index (0.00001).
Specifications
Type NIST Certified Calibration Oil
Scale Range 18.04 Brix
Refractive Index 1.35999
For Use With (Equipment) Refractometer
Includes NIST certification of traceability
Product Identifier
  • NIST Certified Calibration Oil: 1.36000 RI, 50g approx.
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 4
Hazard Statement
  • H413-May cause long lasting harmful effects to aquatic life.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : perfluoro compounds,C5-18