missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ NME1 (Human) Recombinant Protein
Human NME1 full-length ORF ( AAH00293, 1 a.a. - 152 a.a.) recombinant protein with GST-tag at N-terminal.
335.00 € - 508.00 €
Specifications
Accession Number | AAH00293 |
---|---|
Gene ID (Entrez) | 4830 |
Name | non-metastatic cells 1, protein (NM23A) expressed in |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16128401
|
Abnova™
H00004830-P01.10ug |
10 μg |
335.00 €
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16138401
|
Abnova™
H00004830-P01.25ug |
25 μg |
508.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Sequence: MANCERTFIAIKPDGVQRGLVGEIIKRFEQKGFRLVGLKFMQASEDLLKEHYVDLKDRPFFAGLVKYMHSGPVVAMVWEGLNVVKTGRVMLGETNPADSKPGTIRGDFCIQVGRNIIHGSDSVESAEKEIGLWFHPEELVDYTSCAQNWIYE
Specifications
AAH00293 | |
non-metastatic cells 1, protein (NM23A) expressed in | |
125% SDS-PAGE Stained with Coomassie Blue | |
AWD, GAAD, NB, NBS, NDPK-A, NDPKA, NM23, NM23-H1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
4830 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NME1 | |
GST |
Safety and Handling
missing translation for 'shelfLife' : Best use within three months from the date of receipt of this protein