missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ NNMT (Human) Recombinant Protein
Human NNMT full-length ORF ( AAH00234, 1 a.a. - 264 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00004837-P01.25ug
This item is not returnable.
View return policy
Description
- Sequence: MESGFTSKDTYLSHFNPRDYLEKYYKFGSRHSAESQILKHLLKNLFKIFCLDGVKGDLLIDIGSGPTIYQLLSACESFKEIVVTDYSDQNLQELEKWLKKEPEAFDWSPVVTYVCDLEGNRVKGPEKEEKLRQAVKQVLKCDVTQSQPLGAVPLPPADCVLSTLCLDAACPDLPTYCRALRNLGSLLKPGGFLVIMDALKSSYYMIGEQKFSSLPLGREAVEAAVKEAGYTIEWFEVISQSYSSTMANNEGLFSLVARKLSRPL
Specifications
AAH00234 | |
nicotinamide N-methyltransferase | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
4837 | |
Wheat germ expression system | |
25 μg | |
NNMT | |
GST |