Learn More
Novus Biologicals™ Inorganic Pyrophosphatase/PPA1 Protein
Highly purified. Generating reliable and reproducible results.
Brand: Novus Biologicals™ NBP1-72378-100UG
This item is not returnable.
View return policy
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-176 of E.coli Inorganic Pyrophosphatase/PPA1 The Recombinant E. coli Inorganic Pyrophosphatase/PPA1 Protein is derived from E. coli. The Recombinant E. coli Inorganic Pyrophosphatase/PPA1 Protein has been validated for the following applications: SDS-Page.Specifications
SDS-PAGE | |
21.9kDa | |
100μg | |
Store at -80°C. Avoid freeze-thaw cycles. | |
Inorganic Pyrophosphatase/PPA1 | |
>95% |
Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 1mM DTT, 10% glycerol, 50mM NaCl | |
SDS-PAGE | |
Bacteria | |
RUO | |
Unconjugated |