Learn More
Invitrogen™ NPY Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA124891
Description
Recommended positive controls: rat brain. PA1-24891 is expected to cross react with chicken (99% conserved) due to sequence homology. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a neuropeptide that is widely expressed in the central nervous system and influences many physiological processes, including cortical excitability, stress response, food intake, circadian rhythms, and cardiovascular function. The neuropeptide functions through G protein-coupled receptors to inhibit adenylyl cyclase, activate mitogen-activated protein kinase, regulate intracellular calcium levels, and activate potassium channels. A polymorphism in this gene resulting in a change of leucine 7 to proline in the signal peptide is associated with elevated cholesterol levels, higher alcohol consumption, and may be a risk factor for various metabolic and cardiovascular diseases.
Specifications
| NPY | |
| Polyclonal | |
| Unconjugated | |
| NPY | |
| 0710005A05Rik; C-flanking peptide of NPY; CPON; involved in appetite regulation; LOC100912228; Neuropeptide tyrosine; Neuropeptide Y; neuropeptide Y precursor; Neuropeptide-Y; Npy; NPY02; OTTHUMP00000201947; prepro-neuropeptide Y; proneuropeptide Y; pro-neuropeptide Y; pro-neuropeptide Y-like; PYY4; RATNPY; RATNPY02 | |
| Rabbit | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat, Sheep, Pig | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Frozen) | |
| Conc. Not Determined | |
| Whole serum with 15mM sodium azide | |
| P01303, P01304, P07808, P14765, P57774 | |
| NPY | |
| Synthetic neuropeptide Y (NPY, porcine), conjugated to KLH. | |
| RUO | |
| 100912228, 109648, 24604, 397304, 443508, 4852 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.