missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NR2F1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
| Antigen | NR2F1 |
|---|---|
| Concentration | 0.2 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16863055
|
Proteintech
24573-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16853055
|
Proteintech
24573-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene is a nuclear hormone receptor and transcriptional regulator. The encoded protein acts as a homodimer and binds to 5′-AGGTCA-3′ repeats. Defects in this gene are a cause of Bosch-Boonstra optic atrophy syndrome (BBOAS).Specifications
| NR2F1 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| P10589, Q60632 | |
| 13865, 7025 | |
| NR2F1 Fusion Protein Ag0118 | |
| Primary | |
| -20°C | |
| NR2F1 |
| 0.2 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| COUP TF I, COUP TF1, COUP TFI, COUP transcription factor 1, COUP transcription factor I, EAR 3, EAR3, ERBAL3, NR2F1, NR2F2, SVP44, TCFCOUP1, TFCOUP1, V erbA related protein 3 | |
| NR2F1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title