missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NS 3861

Product Code. p-200050933
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18025702

Brand: Tocris Bioscience™ 4831/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

α3β2 full agonist; also α3β4 partial agonist

NS 3861 is a α3β2 full agonist (EC50 = 1.6 μM). Also α3β4 partial agonist (EC50 = 1 μM). Exhibits minimal activity at α4β2 and α4β4. Displays high affinity for α3β4, α4β4, α3β2 and α4β2 (Ki values are 0.62, 7.8, 25 and 55 nM respectively).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(3-Bromo-2-thienyl)-8-methyl-8-azabicyclo[3.2.1]oct-2-ene fumarate
CAS 216853-60-0
Quantity 25 mg
Target Additional Nicotinic Receptor Agonists
Molecular Formula C12H14BrNS.C4H4O4
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.