Learn More
NSC16168, MedChemExpress
MedChemExpress NSC16168 is a specific inhibitor of ERCC1-XPF, with an IC50 value of 0.42 μM. NSC16168 inhibits DNA repair and potentiates CDDP efficacy in cancer.
Brand:  Medchem Express HY-100690
Encompass_Preferred
 View more versions of this product
Product Code. 16454416
  • POA / 100mg
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
NSC16168 is a specific inhibitor of ERCC1-XPF, with an IC50 value of 0.42 μM. NSC16168 inhibits DNA repair and potentiates CDDP efficacy in cancer.
Specifications
Specifications
NSC16168
Light Nude
C17H15NO9S3
DMSO : 31.25 mg/mL (66.00 mM; Need ultrasonic)
473.5
98.0%
Powder
6837-93-0
Research
100 mg
NC1=C2C(C=C(S(=O)(O)=O)C=C2OS(C3=CC=C(C)C=C3)(=O)=O)=CC(S(O)(=O)=O)=C1
473.5
Research
Cancer-programmed cell death