missing translation for 'onlineSavingsMsg'
Learn More

NSE gamma Mouse, Clone: ENO2/1375, enQuireBio™

Mouse Monoclonal Antibody

199.00 € - 510.00 €

Specifications

Antigen NSE gamma
Clone ENO2/1375
Concentration 200μg/mL
Applications Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15939947
View Documents Promotion Details
EnquireBio
2026-MSM1-P0
20 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15949947
View Documents Promotion Details
EnquireBio
2026-MSM1-P1
100 μg
510.00 €
100µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NSE gamma
200μg/mL
Monoclonal
Mouse
10mM PBS with 0.05% BSA and 0.05% Azide
2026
A synthetic peptide corresponding to aa416-433 of human NSE gamma
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
50kDa
ENO2/1375
Immunohistochemistry
Unconjugated
RUO
NP_001966
ENO2
IgG2b
Protein A/G Purified
Cytoplasmic
HepG2, SH-SY-5Y, HeLa or Y79 cells. Pancreas, Cerebellum or Pheochromocytoma.
Videos
SDS
Documents

Documents

Product Certifications