Learn More
WTW™ NT Multirange Universal Power Supply for WTW™ inoLab™
Click to view available options
For Use With (Equipment):
inoLab 340i, 350i, 197i, 1970i Series
Unit Size:
Each
Specifications
Specifications
| Type | Universal Power Supply |
| Electrical Requirements | 100 to 230 V, 50/60 Hz |
| Frequency | 50/60 Hz |
| Voltage | 100 to 240 V |
| For Use With (Equipment) | inoLab 340i, 350i, 197i, 1970i Series |