missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ NUCB1 (Human) Recombinant Protein
Human NUCB1 full-length ORF ( AAH02356, 28 a.a. - 461 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00004924-P01.10ug
This item is not returnable.
View return policy
Description
- Sequence: PLERGAPNKEETPATESPDTGLYYHRYLQEVIDVLETDGHFREKLQAANAEDIKSGKLSRELDFVSHHVRTKLDELERQEVSRPRMLLKAKMDAEQDPNVQVDHLNLLKQFEHLDPQNQHTFEARDLELLIQTATRDLAQYDAAHHEEFKRYEMLKEHERRRYLESLGEEQRKEAERKLEEQQRRHREHPKVNVPGSQAQLKEVWEELDGLDPNRFNPKTFFILHDINSDGVLDEQELEALFTKELEKVYDPKNEEDDMREMEEERLRMREHVMKNVDTNQDRLVTLEEFLASTQRKEFGDTGEGWETVEMHPAYTEEELRRFEEELAAREAELNAKAQRLSQETEALGRSQGRLEAQKRELQQAVLHMEQRKQQQQQQQGHKAPAAHPEGQLKFHPDTDDVPVPAPAGDQKEVDTSEKKLLERLPEVEVPQHL
Specifications
AAH02356 | |
nucleobindin 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NUCB1 | |
GST |
4924 | |
Wheat germ expression system | |
10 μg | |
DKFZp686A15286, FLJ40471, NUC | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |